Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Section lifters

Section lifters

150 mm, 50 mm, 10 mm
Catalog # 231-0293
Supplier:   RSG
Section lifters
Catalog # 231-0293
Supplier:  RSG
Section lifters
Section lifters

150 mm, 50 mm, 10 mm

Price
SFr. 72.90
Log in for availability
1 * 5 ST
Product Details & Documents

Section lifters with flexible blade and handle made of stainless steel.

Documents

Specifications
  • Length:
    150 mm
  • Blade width:
    10 mm
  • Blade length:
    50 mm