Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Section lifters

Section lifters

150 mm, 50 mm, 15 mm
Catalog # 231-0294
Supplier:   RSG
Section lifters
Catalog # 231-0294
Supplier:  RSG
Section lifters
Section lifters

150 mm, 50 mm, 15 mm

Price
SFr. 73.70
Log in for availability
1 * 5 ST
Product Details & Documents

Section lifters with flexible blade and handle made of stainless steel.

Documents

Specifications
  • Length:
    150 mm
  • Blade width:
    15 mm
  • Blade length:
    50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.