Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Section lifters

Section lifters

150 mm, 50 mm, 20 mm
Catalog # RSGA083.020
Supplier:   RSG
Section lifters
Catalog # RSGA083.020
Supplier:  RSG
Section lifters
Section lifters

150 mm, 50 mm, 20 mm

Price
SFr. 74.10
Log in for availability
1 * 5 ST
Product Details & Documents

Section lifters with flexible blade and handle made of stainless steel.

Documents

Specifications
  • Length:
    150 mm
  • Blade width:
    20 mm
  • Blade length:
    50 mm