Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Instrument trays, rectangular

Instrument trays, rectangular

30 mm, 180 mm, 240 mm
Catalog # RSGA701.224
Supplier:   RSG
Instrument trays, rectangular
Catalog # RSGA701.224
Supplier:  RSG
Instrument trays, rectangular
Instrument trays, rectangular

30 mm, 180 mm, 240 mm

Price
SFr. 59.00
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, 18/10.

  • Smooth surfaces for fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance

Documents

Specifications
  • Width:
    180 mm
  • Height:
    30 mm
  • Length:
    240 mm