Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Instrument trays, rectangular

Instrument trays, rectangular

30 mm, 180 mm, 280 mm
Catalog # RSGA701.228
Supplier:   RSG
Instrument trays, rectangular
Catalog # RSGA701.228
Supplier:  RSG
Instrument trays, rectangular
Instrument trays, rectangular

30 mm, 180 mm, 280 mm

Price
SFr. 75.80
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, 18/10.

  • Smooth surfaces for fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance

Documents

Specifications
  • Width:
    180 mm
  • Height:
    30 mm
  • Length:
    280 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.