Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Instrument trays, rectangular

Instrument trays, rectangular

40 mm, 210 mm, 310 mm
Catalog # RSGA701.230
Supplier:   RSG
Instrument trays, rectangular
Catalog # RSGA701.230
Supplier:  RSG
Instrument trays, rectangular
Instrument trays, rectangular

40 mm, 210 mm, 310 mm

Price
SFr. 96.20
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, 18/10.

  • Smooth surfaces for fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance

Documents

Specifications
  • Width:
    210 mm
  • Height:
    40 mm
  • Length:
    310 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.