Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Spoon spatulas, micro

Spoon spatulas, micro

185 mm, 7 mm
Catalog # 231-0470
Supplier:   RSG
Spoon spatulas, micro
Catalog # 231-0470
Supplier:  RSG
Spoon spatulas, micro
Spoon spatulas, micro

185 mm, 7 mm

Price
47.60 SFr.
Log in for availability
1 * 5 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic.

  • With a micro spoon at one end and a micro spatula at the other
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean
Specifications
  • Length:
    185 mm
  • Blade width:
    7 mm