Order Entry
Switzerland
Orders LinkContactUsLinkComponent
StartingBlock™ Blocking Buffer

StartingBlock™ Blocking Buffer

StartingBlock™ (PBS), blocking buffer, 1 L, 1 L
Catalog # PIER37538
StartingBlock™ Blocking Buffer
Catalog # PIER37538
CAS Number:  
StartingBlock™ Blocking Buffer
StartingBlock™ Blocking Buffer

StartingBlock™ (PBS), blocking buffer, 1 L, 1 L

CAS Number:  
Price
SFr. 485.00
Log in for availability
1 * 1 L
Product Details & Documents

These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.

  • Compatible with many detection systems (Western blot, ELISA and IHC)
  • Short blocking times
  • High signal:noise ratio
  • Strip and reprobe without reblocking
  • Available ready to use 1X formulations in PBS and TBS
  • For research use only
  • Not for use in diagnostic procedures

It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock blocking buffer is available in PBS and TBS, with and without Tween 20.

Documents

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    StartingBlock™ (PBS), blocking buffer, 1 L
  • Cat. No.:
    PIER37538
  • Physical form:
    Liquid
  • Concentration:
    1X
  • Storage buffer:
    PBS
  • For use with:
    Western blot, ELISA, Immunohistochemistry (IHC)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.