Orders LinkContactUsLinkComponent
Sample tubes
Sample tubes
2,0 ml, Conical bottom, Clear, with side label, PP, screw cap without attachment loop, Sterile
Katalognr. BZYM710053
Marketsource
Anbieter:   BIOZYM
Sample tubes
Katalognr. BZYM710053
Anbieter:  BIOZYM
Marketsource
Sample tubes
2,0 ml, Conical bottom, Clear, with side label, PP, screw cap without attachment loop, Sterile
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 500 ST
Product Details & Documents
PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
With or without screw caps.
Technische Daten
  • Sterility:
    Sterile
  • Type:
    screw cap without attachment loop
  • Capacity:
    2,0 ml
  • Colour:
    Clear
  • Description:
    with side label
  • Material:
    PP
  • Seq:
    21
  • Base:
    Conical bottom