Sample tubes
1,5 ml, Free standing, Amber, with side label, PP
BZYM710021
Marketsource
: BIOZYM
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
- Capacity:1,5 ml
- Colour:Amber
- Description:with side label
- Material:PP
- Seq:08
- Base:Free standing
- Cat. No.:BZYM710021