Orders LinkContactUsLinkComponent
Sample tubes
1,5 ml, Conical bottom, Amber, with side label, PP
Catalog # BZYM710006
Marketsource
Supplier:   BIOZYM
Sample tubes
Catalog # BZYM710006
Supplier:  BIOZYM
Marketsource
Sample tubes
1,5 ml, Conical bottom, Amber, with side label, PP
Price
Inquire For Price
Log in for availability
1 * 500 ST
Product Details & Documents
PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
With or without screw caps.
Specifications
  • Capacity:
    1,5 ml
  • Colour:
    Amber
  • Description:
    with side label
  • Material:
    PP
  • Seq:
    06
  • Base:
    Conical bottom