Empty solid phase extraction columns, CHROMABOND®
Empty PP columns with PE frits, one filter element is already inserted in the PP column, 45 ml
MANA730355
Empty and reservoir columns with accessories for solid phase extraction.
- Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
- Reservoir columns from PP for application of medium-size samples
- Accessories for SPE columns (filter elements, adapters)
- Capacity:45 ml
- Description:Empty PP columns with PE frits, one filter element is already inserted in the PP column
- Seq:06
Accessories
- Description
- Description
- MANA732020PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weight1 * 250 ST - SFr. 206.00
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description



