Empty solid phase extraction columns, CHROMABOND®
Empty glass columns with glass fiber frits, one filter element is already inserted in the glass column, 6 ml
Catalog # MANA730172
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Empty and reservoir columns with accessories for solid phase extraction.
- Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
- Reservoir columns from PP for application of medium-size samples
- Accessories for SPE columns (filter elements, adapters)
Specifications
- Capacity:6 ml
- Description:Empty glass columns with glass fiber frits, one filter element is already inserted in the glass column
- Seq:10
Accessories
Product Information
- Catalog #AvailabilityPrice
- PE frits for LV PP columns 15 ml, for 100 mg adsorbent weightPE frits for LV PP columns 15 ml, for 100 mg adsorbent weightMANA732019Log in for availability1 * 250 ST - SFr. 147.00
Product Information
- Catalog #AvailabilityPrice
- PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weightPE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weightMANA732020Log in for availability1 * 250 ST - SFr. 206.00
Product Information
- Catalog #AvailabilityPrice
- Adapters (PP) for PP columns (1, 3 and 6 ml)Adapters (PP) for PP columns (1, 3 and 6 ml)MANA730101Log in for availability1 * 10 ST - SFr. 107.00
Product Information
- Catalog #AvailabilityPrice
- Adapters (PVDF) for glass columns (3 and 6 ml)Adapters (PVDF) for glass columns (3 and 6 ml)MANA730105Log in for availability1 * 10 ST - SFr. 223.00
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 70 ml
Product Information
- Catalog #AvailabilityPrice
- Glass fiber frits for glass columns, 6 mlGlass fiber frits for glass columns, 6 mlMANA730192Log in for availability1 * 250 ST - SFr. 265.00
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 15 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 45 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 150 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 30 ml
Product Information
- Catalog #AvailabilityPrice
- Adapters (PE) for PP columns (30 and 70 ml)Adapters (PE) for PP columns (30 and 70 ml)MANA730566Log in for availability1 * 1 KIT - SFr. 32.40
Product Information
- Catalog #AvailabilityPrice
- Adapters (PE) for PP columns (15, 45 and 70 ml)Adapters (PE) for PP columns (15, 45 and 70 ml)MANA730385Log in for availability1 * 10 ST - SFr. 139.00
Product Information
- Catalog #AvailabilityPrice
- Glass fiber frits for glass columns, 3 mlGlass fiber frits for glass columns, 3 mlMANA730191Log in for availability1 * 250 ST - SFr. 218.00
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 1 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 6 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 3 ml
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



