Empty solid phase extraction columns, CHROMABOND®
Empty PP columns with PE frits, one filter element is already inserted in the PP column, 1 ml
MANA730159
Empty and reservoir columns with accessories for solid phase extraction.
- Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
- Reservoir columns from PP for application of medium-size samples
- Accessories for SPE columns (filter elements, adapters)
- Capacity:1 ml
- Description:Empty PP columns with PE frits, one filter element is already inserted in the PP column
- Seq:01
Accessories
- Description
- Description
- MANA732020PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weight1 * 250 ST - SFr. 206.00
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



