Empty solid phase extraction columns, CHROMABOND®
Reservoir columns, PP, with one adapter for 1, 3, 6 ml CHROMABOND® PP columns, 30 ml
Catalog # MANA730103
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Empty and reservoir columns with accessories for solid phase extraction.
- Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
- Reservoir columns from PP for application of medium-size samples
- Accessories for SPE columns (filter elements, adapters)
Specifications
- Capacity:30 ml
- Description:Reservoir columns, PP, with one adapter for 1, 3, 6 ml CHROMABOND® PP columns
- Seq:14
Accessories
Product Information
- Catalog #AvailabilityPrice
- PE frits for LV PP columns 15 ml, for 100 mg adsorbent weightPE frits for LV PP columns 15 ml, for 100 mg adsorbent weightMANA732019Log in for availability1 * 250 ST - SFr. 147.00
Product Information
- Catalog #AvailabilityPrice
- PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weightPE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weightMANA732020Log in for availability1 * 250 ST - SFr. 206.00
Product Information
- Catalog #AvailabilityPrice
- Adapters (PP) for PP columns (1, 3 and 6 ml)Adapters (PP) for PP columns (1, 3 and 6 ml)MANA730101Log in for availability1 * 10 ST - SFr. 107.00
Product Information
- Catalog #AvailabilityPrice
- Adapters (PVDF) for glass columns (3 and 6 ml)Adapters (PVDF) for glass columns (3 and 6 ml)MANA730105Log in for availability1 * 10 ST - SFr. 223.00
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 70 ml
Product Information
- Catalog #AvailabilityPrice
- Glass fiber frits for glass columns, 6 mlGlass fiber frits for glass columns, 6 mlMANA730192Log in for availability1 * 250 ST - SFr. 265.00
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 15 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 45 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 150 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 30 ml
Product Information
- Catalog #AvailabilityPrice
- Adapters (PE) for PP columns (30 and 70 ml)Adapters (PE) for PP columns (30 and 70 ml)MANA730566Log in for availability1 * 1 KIT - SFr. 32.40
Product Information
- Catalog #AvailabilityPrice
- Adapters (PE) for PP columns (15, 45 and 70 ml)Adapters (PE) for PP columns (15, 45 and 70 ml)MANA730385Log in for availability1 * 10 ST - SFr. 139.00
Product Information
- Catalog #AvailabilityPrice
- Glass fiber frits for glass columns, 3 mlGlass fiber frits for glass columns, 3 mlMANA730191Log in for availability1 * 250 ST - SFr. 218.00
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 1 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 6 ml
Product Information
- Catalog #AvailabilityPrice
- PE frits for PP columns, 3 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



