D-(-)-Glutamic acid
Glass bottle for solids, 100 g
Catalog # BACE4030628.0100
Marketsource
Supplier: BACHEM BIOCHEMICA
D-(-)-Glutamic acid
Catalog # BACE4030628.0100
Supplier: BACHEM BIOCHEMICA
Marketsource
CAS Number: 6893-26-1
Jump to:
Product Details & Documents
Synonyms: (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
- CAS Number:6893-26-1
- Formula:C₅H₉NO₄
- MDL Number:MFCD00063112
- Molecular Weight:147.13 g/mol
- Merck Index:13,04482
- Boiling Pt:325 °C (1013 hPa)
- Melting Pt:213 °C
- Density:1.533 g/cm³ (20 °C)
- EINECS:230-000-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



