D-(-)-Glutamic acid
Glass bottle for solids, 25 g
Katalognr. BACE4030628.0025
Marketsource
Anbieter: BACHEM BIOCHEMICA
D-(-)-Glutamic acid
Katalognr. BACE4030628.0025
Anbieter: BACHEM BIOCHEMICA
Marketsource
CAS Nummer: 6893-26-1
Direkt zu
Product Details & Documents
Synonyme: (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
- CAS Nummer:6893-26-1
- Formel:C₅H₉NO₄
- MDL-Nummer:MFCD00063112
- Molekulargewicht:147.13 g/mol
- Merck-Index:13,04482
- Siedepunkt:325 °C (1013 hPa)
- Schmelzender Platingehalt:213 °C
- Dichte:1.533 g/cm³ (20 °C)
- EINECS:230-000-8
Technische Daten
Technische Daten
- Pack type:Glass bottle for solids
- Pk:25 g



