Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

β-Alanine

Glass bottle for solids, 100 g
Catalog # BACE4030329.0100
Marketsource
Supplier:   BACHEM BIOCHEMICA
β-Alanine
Catalog # BACE4030329.0100
Supplier:  BACHEM BIOCHEMICA
Marketsource
CAS Number:  107-95-9
β-Alanine

Glass bottle for solids, 100 g

CAS Number:  107-95-9
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
  • CAS Number:107-95-9
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008200
  • Molecular Weight:89.09 g/mol
  • Merck Index:13,00204
  • Boiling Pt:201 °C (1013 hPa)
  • Melting Pt:94 °C
  • Density:1.44 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-536-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications