Order Entry
Switzerland
Orders LinkContactUsLinkComponent
StartingBlock™ Blocking Buffer

StartingBlock™ Blocking Buffer

StartingBlock™ (TBS), blocking buffer, 1 L, 1 L
Catalog # PIER37542
StartingBlock™ Blocking Buffer
Catalog # PIER37542
CAS Number:  
StartingBlock™ Blocking Buffer
StartingBlock™ Blocking Buffer

StartingBlock™ (TBS), blocking buffer, 1 L, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents

These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.

  • Compatible with many detection systems (Western blot, ELISA and IHC)
  • Short blocking times
  • High signal:noise ratio
  • Strip and reprobe without reblocking
  • Available ready to use 1X formulations in PBS and TBS
  • For research use only
  • Not for use in diagnostic procedures

It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock blocking buffer is available in PBS and TBS, with and without Tween 20.

Documents

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    StartingBlock™ (TBS), blocking buffer, 1 L
  • Cat. No.:
    PIER37542
  • Physical form:
    Liquid
  • Concentration:
    1X
  • Storage buffer:
    TBS
  • For use with:
    Western blot, ELISA, Immunohistochemistry (IHC)

Specifications