Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Scoops

Scoops

310 mm, 110 mm, White
Catalog # SCRW4280310
Supplier:   SCHROEDER WILESCO
Scoops
Catalog # SCRW4280310
Supplier:  SCHROEDER WILESCO
Scoops
Scoops

310 mm, 110 mm, White

Price
SFr. 4.40
Log in for availability
1 * 1 ST
Product Details & Documents

These coloured scoops made from PP are suitable for food contact.

Specifications
  • Width:
    110 mm
  • Colour:
    White
  • Length:
    310 mm
  • Seq:
    02