Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Scoops

Scoops

210 mm, 75 mm, White
Catalog # SCRW4280210
Supplier:   SCHROEDER WILESCO
Scoops
Catalog # SCRW4280210
Supplier:  SCHROEDER WILESCO
Scoops
Scoops

210 mm, 75 mm, White

Price
3.50 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

These coloured scoops made from PP are suitable for food contact.

Specifications
  • Width:
    75 mm
  • Colour:
    White
  • Length:
    210 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.