Orders LinkContactUsLinkComponent
Forceps
Forceps
Straight, blunt, 200 mm
Catalog # 232-0033
Supplier:   MURRAY S
Forceps
Catalog # 232-0033
Supplier:  MURRAY S
Forceps
Forceps
Straight, blunt, 200 mm
Price
SFr. 21.40
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Blunt, narrow flutes
  • For dissecting and general applications
Specifications
  • Type:
    Straight, blunt
  • Length:
    200 mm