Buffer solutions, Certipur®, colour coded, Supelco®
Catalog # 1.09400.9010
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Materials for the precise calibration and monitoring of pH meters.
- Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
- Colour coded solutions for quick identification
- Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
- Reference temperature: +20 °C
Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).
Documents
Certificates
Search by Lot NumberCertipur Merck Buffer Calibration Standards for pH Measurements
Product Literature
ProductPage_109475 ProductPage_109476 ProductPage_109477 Spec MERC1.09400.0500.pdf
Product specification
Specifications
Specifications
- Pack type:Titripac®
- pH buffer type:Borate
- Pk:10 L
- Description:Certipur® buffer solution, pH 10,00, yellow
- Cat. No.:1.09400.9010
- M-Clarity quality level:MQ300
- Environmentally Preferable:Sustainable Packaging
Specifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



