Buffer solutions, Certipur®, colour coded, Supelco®
Certipur® buffer solution, pH 10,00, yellow, Borate, Plastic bottle, 500 mL
Anbieter: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Katalognr. 1.09400.0500
Anbieter: Merck
CAS Nummer:
Direkt zu
Product Details & Documents
Materials for the precise calibration and monitoring of pH meters.
- Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
- Colour coded solutions for quick identification
- Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
- Reference temperature: +20 °C
Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).
Dokumente
Zertifikate
Suche nach ChargennummerCertipur Merck Buffer Calibration Standards for pH Measurements
Produktliteratur
MERC1.09400.0500_TDS_EN TDS Certified Reference Materials
Technisches Datenblatt
MERC1.09400.4000 msds_109400 msds_109475 msds_109475 msds_109476
Sicherheitsdatenblatt (SDS)
Mehr
ProductPage_109475 ProductPage_109476 ProductPage_109477 Spec MERC1.09400.0500.pdf
Produktspezifikation
Technische Daten
Technische Daten
- Pack type:Plastic bottle
- pH buffer type:Borate
- Pk:500 mL
- Description:Certipur® buffer solution, pH 10,00, yellow
- Cat. No.:1.09400.0500
- M-Clarity quality level:MQ300
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



