Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
500 ml, 315 mm
Catalog # 231-0120
Supplier:   Brand
Measuring scoops
Catalog # 231-0120
Supplier:  Brand
Measuring scoops
Measuring scoops
500 ml, 315 mm
Price
SFr. 44.90
Log in for availability
1 * 6 ST
Product Details & Documents
PP, white. With flat, bevelled end and reinforced handle.
  • Excellent chemical resistance, unbreakable
  • The capacity of the scoop is stamped into the handle for easy reading
Specifications
  • Capacity:
    500 ml
  • Length:
    315 mm