Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Attachable screw thread tubes

Attachable screw thread tubes

GL 32, 28 mm, 140 mm
Catalog # 201-1607
Supplier:   Lenz Laborglas
Attachable screw thread tubes
Catalog # 201-1607
Supplier:  Lenz Laborglas
Attachable screw thread tubes
Attachable screw thread tubes

GL 32, 28 mm, 140 mm

Price
9.40 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass, clear.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

DIN screw thread GL.

Documents

Specifications
  • Thread:
    GL 32
  • Tube ext.Ø:
    28 mm
  • Seq:
    04
  • Total length:
    140 mm
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..