ContactUsLinkComponent
CONOX-10,COMBINED OXYGEN-/CONDUCTIVITY SENSOR WITH CABLE 33 FT (10 M)WITH 3 EXCHANGE MEMBRANE HEADS, CLEANING SOLUTION, CONDUCTIVITY STANDARD (1413 µS/CM), ABRASIVE FOIL, OXICAL® CX, CALIBRATION VESSEL ELECTROLYTE 1 * 1 items
Catalog # 665-0102
Supplier:  WTW
CONOX-10,COMBINED OXYGEN-/CONDUCTIVITY SENSOR WITH CABLE 33 FT (10 M)WITH 3 EXCHANGE MEMBRANE HEADS, CLEANING SOLUTION, CONDUCTIVITY STANDARD (1413 µS/CM), ABRASIVE FOIL, OXICAL® CX, CALIBRATION VESSEL ELECTROLYTE 1 * 1 items
Catalog # 665-0102
Supplier:  WTW
Frequently Bought Together