ContactUsLinkComponent
CONOX-6,COMBINED OXYGEN-/CONDUCTIVITY SENSOR WITH CABLE 19,8 FT (6 M)WITH 3 EXCHANGE MEMBRANE HEADS, CLEANING SOLUTION, CONDUCTIVITY STANDARD (1413 µS/CM), ABRASIVE FOIL, OXICAL® CX, CALIBRATION VESSEL ELECTROLYTE 1 * 1 items
Catalog # 665-0101
Supplier:  WTW
CONOX-6,COMBINED OXYGEN-/CONDUCTIVITY SENSOR WITH CABLE 19,8 FT (6 M)WITH 3 EXCHANGE MEMBRANE HEADS, CLEANING SOLUTION, CONDUCTIVITY STANDARD (1413 µS/CM), ABRASIVE FOIL, OXICAL® CX, CALIBRATION VESSEL ELECTROLYTE 1 * 1 items
Catalog # 665-0101
Supplier:  WTW
Frequently Bought Together