Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Spray bottles, WHEATON®

Spray bottles, WHEATON®

35 ml, 94 mm, with PP screw cap
Catalog # WHEA226434
Supplier:   DWK Life Sciences
Spray bottles, WHEATON®
Catalog # WHEA226434
Supplier:  DWK Life Sciences
Spray bottles, WHEATON®

35 ml, 94 mm, with PP screw cap

Price
SFr. 563.00
Log in for availability
1 * 144 ST
Product Details & Documents

Bottle made of white LDPE (Low Density Polyethylene).

  • Includes bottle, atomizer tip, dip tube and screw cap
  • Screw cap made from polypropylene

Oval spray bottle.

Specifications
  • Type:
    with PP screw cap
  • Capacity:
    35 ml
  • Height:
    94 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.