Replacement impeller for Magna Flex™ flasks
Replacement impeller for 250 ml Magna Flex® spinner flask
Catalog # 734-3144
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
This unit includes a top cap with liners, glass stirring ball and connecting silicone tubing. This glass impeller assembly is designed for replacement on all Magna Flex™ style flasks.
Specifications
- Description:Replacement impeller for 250 ml Magna Flex® spinner flask
- Seq:02