Diagnostic vials, Nalgene®
Product Details & Documents
These clear PETG vials with white HDPE closures are ideal for sampling, storage and shipment of reagents or buffer solutions.
- Non-pyrogenic and non-cytotoxic
- Sterile to 10⁻⁶ SAL
- 20-415 lined white high-density polyethylene closure attached to PETG vial
- Leakproof when used with Nalgene closures
- Narrow mouth
These bottles are constructed from non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white HDPE (high-density polyethylene) closures.
Inner neck diameter: 11 mm
Certifications: Manufactured in compliance with ISO 13485:2003 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Packaging: Packed in shrink-wrapped trays with closures attached.
Specifications
- Cap included:Yes
- Cap Size:20-415
- Ø×H:30×66 mm
- Sterile:+
- Capacity:20,00 ml
- Seq:03
- Cat. No.:391-0076
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - Environmentally Preferable:Low Manufacturing Impact
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



