Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Microsette™ M503

Biopsy processing/embedding cassettes, Microsette™ M503

Beige
Catalog # 720-1029
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Microsette™ M503
Catalog # 720-1029
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Microsette™ M503
Biopsy processing/embedding cassettes, Microsette™ M503

Beige

Price
623.00 SFr.
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer.

  • No need for biopsy pads
  • Six compartments, each 7×12 mm, numbered 1 to 6
  • 2000 square openings (each 0,26 mm) to maximise fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Specifications
  • Colour:
    Beige
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.