Tissue processing/embedding cassettes, Histosette® II, M485
Purple
Codice catalogo 720-0223
Nome del fornitore: Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M485
Codice catalogo 720-0223
Nome del fornitore: Simport Scientific
Vai a
Product Details & Documents
Acetal polymer, similar to M492, but covers are already removed from cassettes and are packed separately in the case.
- Convenient for cassette labelling instruments as the covers are already removed
- Resistant to the chemical action of histological solvents
- Flow-through slots maximise fluid exchange
Specifiche
- Colour:Purple
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



