Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Adhesion slides

Adhesion slides

Adhesion slides with 12 wells (Ø = 5 mm)
Katalognr. MARI0900100
Anbieter:   MARIENFELD
Adhesion slides
Katalognr. MARI0900100
Anbieter:  MARIENFELD
Adhesion slides
Adhesion slides

Adhesion slides with 12 wells (Ø = 5 mm)

Preis
SFr. 2’770.00
Einloggen für Verfügbarkeit
1 * 100 ST
Product Details & Documents

Adhesion slides with frosted ends and 90° ground edges.

Technische Daten
  • Description:
    Adhesion slides with 12 wells (Ø = 5 mm)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.