Ammonia solution 25%
1 KIT
Catalog # EURLEMR272001
Supplier: EUROCLONE
Jump to:
Product Details & Documents
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Boiling Pt:32…37.7 °C (1013 hPa)
- Melting Pt:–60…–57.5 °C
- Density:0.88…0.91 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:2672
- ADR:8,III
Specifications
Specifications
- Pk:1 KIT



