Buffer, TE solution (TRIS-EDTA buffer) pH 8,0
TE buffer solution (TRIS-EDTA buffer) pH 8,0, 1 KIT
Catalog # EURLEMR227001
Supplier: EUROCLONE
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0
Catalog # EURLEMR227001
Supplier: EUROCLONE
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 KIT
- Description:TE buffer solution (TRIS-EDTA buffer) pH 8,0



