Order Entry
Switzerland
Orders LinkContactUsLinkComponent
2-Mercaptoethanol

2-Mercaptoethanol

Glass bottle, 250 mL
Catalog # EURLEMR216250
Toxic
Corrosive
Environment
Supplier:   EUROCLONE
2-Mercaptoethanol
Catalog # EURLEMR216250
Supplier:  EUROCLONE
CAS Number:  60-24-2
2-Mercaptoethanol
2-Mercaptoethanol

Glass bottle, 250 mL

CAS Number:  60-24-2
Price
Inquire For Price
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  2-Hydroxyethyl mercaptan, β-Mercaptoethanol
  • CAS Number:60-24-2
  • Formula:HSCH₂CH₂OH
  • MDL Number:MFCD00004890
  • Molecular Weight:78.14 g/mol
  • Merck Index:13,05894
  • Boiling Pt:157 °C (1013 hPa)
  • Melting Pt:–100 °C
  • Density:1.12 g/cm³ (20 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:200-464-6
  • UN:2966
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.