G418
1 KIT
Catalog # EURLEMR143005
Supplier: EUROCLONE
Jump to:
Product Details & Documents
Synonyms: G-418, (2R,3S,4R,5R,6S)-5-Amino-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-2-((R)-1-hydroxyethyl)tetrahydro-2H-pyran-3,4-diol
- CAS Number:49863-47-0
- Formula:C₂₀H₄₀N₄O₁₀
- MDL Number:MFCD30541267
- Molecular Weight:496.552 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pk:1 KIT



