Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Formaldehyde 37% in aqueous solution

Glass bottle, 500 mL
  EURLEMR140500
 
 
 
 :   EUROCLONE
Formaldehyde 37% in aqueous solution
  EURLEMR140500
 :  EUROCLONE
 :  50-00-0
Formaldehyde 37% in aqueous solution

Glass bottle, 500 mL

 :  50-00-0
 
 
   
1 * 500 mL
 
 :  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  •  :50-00-0
  •  :CH₂O
  •  :MFCD00003274
  •  :30.03 g/mol
  •  :Ambient
  •  :200-001-8
  •  :2209
  •  :8,III
 

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL