Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Formaldehyde 37% in aqueous solution

Formaldehyde 37% in aqueous solution

Glass bottle, 250 mL
Catalog # EURLEMR140250
Toxic
Corrosive
Health
Supplier:   EUROCLONE
Formaldehyde 37% in aqueous solution
Catalog # EURLEMR140250
Supplier:  EUROCLONE
CAS Number:  50-00-0
Formaldehyde 37% in aqueous solution

Glass bottle, 250 mL

CAS Number:  50-00-0
Price
Inquire For Price
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:2209
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.