Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Formaldehyde 37% in aqueous solution

Formaldehyde 37% in aqueous solution

1 KIT
Catalog # EURLEMR140001
Toxic
Corrosive
Health
Supplier:   EUROCLONE
Formaldehyde 37% in aqueous solution
Catalog # EURLEMR140001
Supplier:  EUROCLONE
CAS Number:  50-00-0
Formaldehyde 37% in aqueous solution

1 KIT

CAS Number:  50-00-0
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:2209
  • ADR:8,III
Specifications

Specifications

  • Pk:
    1 KIT

Specifications