Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ethidium bromide

Ethidium bromide

1 KIT
Catalog # EURLEMR135010
Toxic
Health
Supplier:   EUROCLONE
Ethidium bromide
Catalog # EURLEMR135010
Supplier:  EUROCLONE
CAS Number:  1239-45-8
Ethidium bromide

1 KIT

CAS Number:  1239-45-8
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Merck Index:13,04751
  • Boiling Pt:72 °C (0.1 mmHg)
  • Melting Pt:261…264 °C
  • Density:1.01 g/cm³ (20 °C)
  • Flash Pt:<100 °C
  • Storage Temperature:Ambient
  • EINECS:214-984-6
  • UN:2811
  • ADR:6.1,I
Specifications

Specifications

  • Pk:
    1 KIT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.