Ethidium bromide
Glass bottle for solids, 1 g
Katalognr. EURLEMR135001
Anbieter: EUROCLONE
Direkt zu
Product Details & Documents
Synonyme: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
- CAS Nummer:1239-45-8
- Formel:C₂₁H₂₀BrN₃
- MDL-Nummer:MFCD00011724
- Molekulargewicht:394.31 g/mol
- Merck-Index:13,04751
- Siedepunkt:72 °C (0.1 mmHg)
- Schmelzender Platingehalt:261…264 °C
- Dichte:1.01 g/cm³ (20 °C)
- Blitz Pt:<100 °C
- Lagertemperatur:Ambient
- EINECS:214-984-6
- UN:2811
- ADR:6.1,I
Technische Daten
Technische Daten
- Pack type:Glass bottle for solids
- Pk:1 g
- Cat. No.:EURLEMR135001
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



