Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-(+)-Mannose

D-(+)-Mannose

1 KIT
Catalog # EURLEMR118100
Supplier:   EUROCLONE
D-(+)-Mannose
Catalog # EURLEMR118100
Supplier:  EUROCLONE
CAS Number:  3458-28-4
D-(+)-Mannose

1 KIT

CAS Number:  3458-28-4
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,05772
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:133 °C
  • Density:1.539 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:222-392-4
Specifications

Specifications

  • Pk:
    1 KIT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.