Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Agar

Agar

1 KIT
Catalog # EURLEMR083500
Supplier:   EUROCLONE
Agar
Catalog # EURLEMR083500
Supplier:  EUROCLONE
CAS Number:  9002-18-0
Agar
CAS Number:  9002-18-0
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specifications

  • Pk:
    1 KIT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.