Actinomycin D
Glass bottle for solids, 5 mg
Catalog # EURLEMR082005
Supplier: EUROCLONE
Jump to:
Product Details & Documents
- CAS Number:50-76-0
- Formula:C₆₂H₈₆N₁₂O₁₆
- MDL Number:MFCD00005033
- Molecular Weight:1255.5 g/mol
- UN:3462
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg



