Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), 1 KIT
Catalog # EURLEMR064001
Supplier:   EUROCLONE
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # EURLEMR064001
Supplier:  EUROCLONE
CAS Number:  
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), 1 KIT

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)

Specifications