Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 500 mL
Catalog # EURLEMR063500
Supplier:   EUROCLONE
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Catalog # EURLEMR063500
Supplier:  EUROCLONE
CAS Number:  
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 500 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer)

Specifications