Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), 1 KIT
Catalog # EURLEMR063001
Supplier: EUROCLONE
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Catalog # EURLEMR063001
Supplier: EUROCLONE
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 KIT
- Description:TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



