Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)
EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 500 mL
Catalog # EURLEMR034500
Supplier: EUROCLONE
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)
Catalog # EURLEMR034500
Supplier: EUROCLONE
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Description:EDTA buffer solution pH 8,0 (0,5 mol/l)



